Learn More
MSC Carlisle Round Waste Bin Trash Container Lid, 10 Gallon, Polyethylene, Purple, NSF Certified
Catalog No. 502839230
Encompass
Bronco™ Round Waste Bin Trash Container Lids are designed for durable, long-lasting performance in various settings. Made from extra-thick polyethylene, these lids feature a design that prevents water collection and are NSF 2 and NSF 21 certified for food safety.
  • Locking lids snap down and press-pull lids stay on and remove with ease.
  • Durable construction made of extra thick plastic for maximum longevity.
  • Food safe: NSF 2 and NSF 21 certified for storing, separating, and color-coding various food products.
  • Design prevents water from collecting on the lid.
  • Color code program aligns with HACCP practices and promotes an integrated color-coded sanitary maintenance system.
  • Recyclable: "2" signifies high-density polyethylene (HDPE).
Catalog No. 50-283-9230 Supplier MSC Supplier No. 73918435
Bronco™ Round Waste Bin Trash Container Lids are designed for durable, long-lasting performance in various settings. Made from extra-thick polyethylene, these lids feature a design that prevents water collection and are NSF 2 and NSF 21 certified for food safety.
  • Locking lids snap down and press-pull lids stay on and remove with ease.
  • Durable construction made of extra thick plastic for maximum longevity.
  • Food safe: NSF 2 and NSF 21 certified for storing, separating, and color-coding various food products.
  • Design prevents water from collecting on the lid.
  • Color code program aligns with HACCP practices and promotes an integrated color-coded sanitary maintenance system.
  • Recyclable: "2" signifies high-density polyethylene (HDPE).