Learn More
MSC Carlisle Trash Container Lid, 55 gallon, Round, HDPE, Blue
Catalog No. 502836034
Encompass
This lid is designed for a 55 gallon round waste bin trash container. Made from durable HDPE, it features a blue color and is part of a color-coded sanitary maintenance system.
  • Locking and press-pull mechanisms for secure attachment and easy removal.
  • Durable construction with extra thick plastic for maximum longevity.
  • NSF 2 and NSF 21 certified for food storage, separation, and color-coding.
  • Design prevents water accumulation on the lid.
  • Aligns with HACCP practices for integrated color-coded sanitary maintenance.
  • NSF listed, ensuring compliance with public health, safety, and environmental protection standards.
  • Made in the USA.
  • Recyclable, made from high-density polyethylene (HDPE).
Catalog No. 50-283-6034 Supplier MSC Supplier No. 73923062
This lid is designed for a 55 gallon round waste bin trash container. Made from durable HDPE, it features a blue color and is part of a color-coded sanitary maintenance system.
  • Locking and press-pull mechanisms for secure attachment and easy removal.
  • Durable construction with extra thick plastic for maximum longevity.
  • NSF 2 and NSF 21 certified for food storage, separation, and color-coding.
  • Design prevents water accumulation on the lid.
  • Aligns with HACCP practices for integrated color-coded sanitary maintenance.
  • NSF listed, ensuring compliance with public health, safety, and environmental protection standards.
  • Made in the USA.
  • Recyclable, made from high-density polyethylene (HDPE).