Learn More
MSC Carlisle Round Waste Bin Lid, 44 Gallon, Red, Polyethylene
Catalog No. 502839622
Encompass
These round waste bin lids are designed for durable waste management in various environments. Made from extra-thick high-density polyethylene (HDPE), this 44-gallon lid in red features a design that prevents water collection and is NSF 2 and NSF 21 certified for food safety, promoting an integrated color-coded sanitary maintenance system.
  • Locking lids snap down and press-pull lids stay on and remove with ease.
  • Durable construction with extra thick plastic ensures maximum longevity.
  • Food safe, NSF 2 and NSF 21 certified for storing, separating, and color-coding various food products.
  • Design prevents water from collecting on the lid.
  • Color code program aligns with HACCP practices for sanitary maintenance.
  • NSF listed, demonstrating commitment to public health, safety, and environmental protection.
  • Made in the USA.
  • Recyclable (high-density polyethylene, HDPE).
Catalog No. 50-283-9622 Supplier MSC Supplier No. 73922064
These round waste bin lids are designed for durable waste management in various environments. Made from extra-thick high-density polyethylene (HDPE), this 44-gallon lid in red features a design that prevents water collection and is NSF 2 and NSF 21 certified for food safety, promoting an integrated color-coded sanitary maintenance system.
  • Locking lids snap down and press-pull lids stay on and remove with ease.
  • Durable construction with extra thick plastic ensures maximum longevity.
  • Food safe, NSF 2 and NSF 21 certified for storing, separating, and color-coding various food products.
  • Design prevents water from collecting on the lid.
  • Color code program aligns with HACCP practices for sanitary maintenance.
  • NSF listed, demonstrating commitment to public health, safety, and environmental protection.
  • Made in the USA.
  • Recyclable (high-density polyethylene, HDPE).