Learn More
MSC Carlisle Round Waste Bin Lid, 10 Gallon, Flat, Polyethylene, Gray
Catalog No. 502839390
Encompass
This heavy-duty lid for waste containers is made from extra-thick polyethylene, offering excellent resistance to acids, bases, and vegetable oils. It features snap-down tabs for a secure seal and a pull release for easy removal. Rounded edges and a textured surface resist abrasions and scuffs, and it withstands temperatures from -50 to 120 degrees Fahrenheit.
  • Keeps waste covered and unpleasant odors at bay
  • Extra-thick polyethylene construction for durability
  • Snap-down tabs provide a secure seal
  • Pull release ensures simple removal
  • Rounded edges and textured surface resist abrasions and scuffs
  • Meets food contact, cleanability, and thermoplastic refuse standards
  • NSF 2 and NSF 21 certified for storing, separating and color-coding various food products
  • Design prevents water from collecting on the lid
  • Part of a color code program aligning with HACCP practices
Catalog No. 50-283-9390 Supplier MSC Supplier No. 73918807
This heavy-duty lid for waste containers is made from extra-thick polyethylene, offering excellent resistance to acids, bases, and vegetable oils. It features snap-down tabs for a secure seal and a pull release for easy removal. Rounded edges and a textured surface resist abrasions and scuffs, and it withstands temperatures from -50 to 120 degrees Fahrenheit.
  • Keeps waste covered and unpleasant odors at bay
  • Extra-thick polyethylene construction for durability
  • Snap-down tabs provide a secure seal
  • Pull release ensures simple removal
  • Rounded edges and textured surface resist abrasions and scuffs
  • Meets food contact, cleanability, and thermoplastic refuse standards
  • NSF 2 and NSF 21 certified for storing, separating and color-coding various food products
  • Design prevents water from collecting on the lid
  • Part of a color code program aligning with HACCP practices