Learn More
MSC Carlisle Round Waste Bin Trash Container Lid, Flat, 44 Gallon, Extra Thick Plastic
Catalog No. 502838694
Encompass
This round waste bin lid is constructed from extra thick plastic for maximum durability in commercial environments. It is designed to snap down easily onto 44-gallon waste containers, facilitating secure closure and easy removal.
  • Designed for commercial environments
  • Made from extra thick plastic for enhanced durability
  • NSF 2 certified for food safety applications
  • Features an easy-to-use snap-down design
  • Suitable for storage, separation, and color-coding of various products
  • Compatible with 44-gallon waste containers
  • Operates within a temperature range of -50 °F (-45.56 °C) to 120 °F (48.89 °C)
Catalog No. 50-283-8694 Supplier MSC Supplier No. 73922346
This round waste bin lid is constructed from extra thick plastic for maximum durability in commercial environments. It is designed to snap down easily onto 44-gallon waste containers, facilitating secure closure and easy removal.
  • Designed for commercial environments
  • Made from extra thick plastic for enhanced durability
  • NSF 2 certified for food safety applications
  • Features an easy-to-use snap-down design
  • Suitable for storage, separation, and color-coding of various products
  • Compatible with 44-gallon waste containers
  • Operates within a temperature range of -50 °F (-45.56 °C) to 120 °F (48.89 °C)