Learn More
MSC Carlisle Round Waste Bin Lid, 55 Gallon, White, Polyethylene, Flat
Catalog No. 502839624
Encompass
This round waste bin lid is designed for 55 gallon containers. Constructed from extra-thick polyethylene, it provides durability and features a snap-lock mechanism to keep contents secure. Its design prevents water collection, and it is certified for food contact, making it suitable for various applications.
  • Snap down tabs lock securely.
  • Extra thick plastic for maximum longevity.
  • Design prevents water from collecting on the lid.
  • Certified for storing, separating, and color-coding various food products.
  • Made in the USA.
  • Recyclable material.
  • Resists acids, bases, and vegetable oils.
  • Withstands temperatures from -50 to 120 degrees Fahrenheit.
  • Rounded edges and textured surface resist abrasions and scuffs.
  • Raised inner circle matches container bases for quick stacking.
Catalog No. 50-283-9624 Supplier MSC Supplier No. 73922577
This round waste bin lid is designed for 55 gallon containers. Constructed from extra-thick polyethylene, it provides durability and features a snap-lock mechanism to keep contents secure. Its design prevents water collection, and it is certified for food contact, making it suitable for various applications.
  • Snap down tabs lock securely.
  • Extra thick plastic for maximum longevity.
  • Design prevents water from collecting on the lid.
  • Certified for storing, separating, and color-coding various food products.
  • Made in the USA.
  • Recyclable material.
  • Resists acids, bases, and vegetable oils.
  • Withstands temperatures from -50 to 120 degrees Fahrenheit.
  • Rounded edges and textured surface resist abrasions and scuffs.
  • Raised inner circle matches container bases for quick stacking.