Learn More
MSC Carlisle Round Trash Container Lid, 20 Gallon, Flat, Green, HDPE
Catalog No. 502836039
Encompass
This lid is designed for 20-gallon waste bins, offering durable construction and secure sealing for various environments. It features a flat design and is certified for food contact, cleanability, and thermoplastic refuse standards.
  • Locking lids snap down for secure sealing and remove with ease.
  • Durable, extra-thick plastic construction for maximum longevity.
  • Food safe, NSF 2 and NSF 21 certified for storing and separating food products.
  • Design prevents water from collecting on the lid.
  • Resistant to acids, bases, and vegetable oils.
  • Withstands temperatures from -50 to 120 degrees Fahrenheit.
  • Rounded edges and textured surface resist abrasions and scuffs.
  • Raised inner circle allows for quick stacking with compatible waste container bases.
Catalog No. 50-283-6039 Supplier MSC Supplier No. 73919441
This lid is designed for 20-gallon waste bins, offering durable construction and secure sealing for various environments. It features a flat design and is certified for food contact, cleanability, and thermoplastic refuse standards.
  • Locking lids snap down for secure sealing and remove with ease.
  • Durable, extra-thick plastic construction for maximum longevity.
  • Food safe, NSF 2 and NSF 21 certified for storing and separating food products.
  • Design prevents water from collecting on the lid.
  • Resistant to acids, bases, and vegetable oils.
  • Withstands temperatures from -50 to 120 degrees Fahrenheit.
  • Rounded edges and textured surface resist abrasions and scuffs.
  • Raised inner circle allows for quick stacking with compatible waste container bases.