Learn More
MSC Carlisle Round Waste Bin Lid, 55 Gallon, Polyethylene, Green
Catalog No. 502835889
Encompass
This round waste bin lid is designed for 55-gallon waste bins, constructed from durable polyethylene (HDPE) in a green color. It features a design that prevents water collection and is food safe, being NSF 2 and NSF 21 certified, making it suitable for various waste management and food storage applications.
  • Locking lids snap down and press-pull lids stay on and remove with ease.
  • Made of extra thick plastic for maximum longevity.
  • NSF 2 and NSF 21 certified for storing, separating, and color-coding various food products.
  • Design prevents water from collecting on the lid.
  • Aligns with HACCP practices and promotes an integrated color-coded sanitary maintenance system.
  • Made in the USA.
  • Recyclable (HDPE).
Catalog No. 50-283-5889 Supplier MSC Supplier No. 73923070
This round waste bin lid is designed for 55-gallon waste bins, constructed from durable polyethylene (HDPE) in a green color. It features a design that prevents water collection and is food safe, being NSF 2 and NSF 21 certified, making it suitable for various waste management and food storage applications.
  • Locking lids snap down and press-pull lids stay on and remove with ease.
  • Made of extra thick plastic for maximum longevity.
  • NSF 2 and NSF 21 certified for storing, separating, and color-coding various food products.
  • Design prevents water from collecting on the lid.
  • Aligns with HACCP practices and promotes an integrated color-coded sanitary maintenance system.
  • Made in the USA.
  • Recyclable (HDPE).