Learn More
MSC Carlisle Recycle Container, Round, 32 Gallon, Polyethylene, Blue
Catalog No. 502835887
Encompass
This durable plastic round recycle container features reinforced drag skids for longevity and a stackable design to prevent vacuum seals. Ergonomic handholds and integrated bag cinches simplify emptying. It is Food Safe, NSF 2 and NSF 21 certified, suitable for color-coded food product management, and aligns with HACCP practices.
  • Durable plastic construction with reinforced drag skids.
  • Stackable design prevents vacuum seal during transport.
  • Ergonomic handholds and integrated bag cinches for easy emptying.
  • Food Safe, NSF 2 and NSF 21 certified.
  • Aligns with HACCP for color-coded sanitary maintenance.
  • Made in the USA.
  • Recyclable (LDPE).
Catalog No. 50-283-5887 Supplier MSC Supplier No. 73996795
This durable plastic round recycle container features reinforced drag skids for longevity and a stackable design to prevent vacuum seals. Ergonomic handholds and integrated bag cinches simplify emptying. It is Food Safe, NSF 2 and NSF 21 certified, suitable for color-coded food product management, and aligns with HACCP practices.
  • Durable plastic construction with reinforced drag skids.
  • Stackable design prevents vacuum seal during transport.
  • Ergonomic handholds and integrated bag cinches for easy emptying.
  • Food Safe, NSF 2 and NSF 21 certified.
  • Aligns with HACCP for color-coded sanitary maintenance.
  • Made in the USA.
  • Recyclable (LDPE).