Learn More
MSC Carlisle Round Waste Bin Trash Container, 20 Gallon, Black, Polyethylene
Catalog No. 502837530
Encompass
This durable round waste bin trash container, constructed from polyethylene, features reinforced drag skids for enhanced longevity and easy movement. Its stackable design prevents vacuum sealing during transport, while ergonomic handholds ensure efficient emptying. Integrated bag cinches are included. It is NSF 2 and NSF 21 certified, suitable for food product storage and separation.
  • Durable plastic construction with reinforced drag skids for maximum longevity
  • Stackable design eliminates vacuum seal during transport
  • Ergonomic hand holds on the bottom for easy and efficient emptying
  • Integrated bag cinches secure liners
  • NSF 2 and NSF 21 certified for safe food product storage and color-coding
  • Color code program aligns with HACCP practices for sanitary maintenance
  • Manufactured in the USA
  • Made from recyclable low-density polyethylene (LDPE)
Catalog No. 50-283-7530 Supplier MSC Supplier No. 73994774
This durable round waste bin trash container, constructed from polyethylene, features reinforced drag skids for enhanced longevity and easy movement. Its stackable design prevents vacuum sealing during transport, while ergonomic handholds ensure efficient emptying. Integrated bag cinches are included. It is NSF 2 and NSF 21 certified, suitable for food product storage and separation.
  • Durable plastic construction with reinforced drag skids for maximum longevity
  • Stackable design eliminates vacuum seal during transport
  • Ergonomic hand holds on the bottom for easy and efficient emptying
  • Integrated bag cinches secure liners
  • NSF 2 and NSF 21 certified for safe food product storage and color-coding
  • Color code program aligns with HACCP practices for sanitary maintenance
  • Manufactured in the USA
  • Made from recyclable low-density polyethylene (LDPE)