Learn More
MSC Carlisle Round Waste Bin Trash Container, 55 Gallon, Green, Polyethylene, NSF Certified
Catalog No. 502837881
Encompass
This 55-gallon round waste bin trash container is designed for durability and efficient waste management. Made from polyethylene, it features a stackable design and ergonomic handles for easy emptying. It is also NSF certified for food safety applications.
  • Made of durable plastic, reinforced with drag skids for maximum longevity.
  • Stackable design prevents vacuum seal during transport.
  • Ergonomic hand holds on the bottom facilitate easy emptying.
  • Integrated bag cinches secure liners.
  • Food safe and NSF 2 and NSF 21 certified for storing food products.
  • Color code program aligns with HACCP practices for sanitary maintenance.
  • Made in the USA.
  • Recyclable (LDPE, code 4).
Catalog No. 50-283-7881 Supplier MSC Supplier No. 73998437
This 55-gallon round waste bin trash container is designed for durability and efficient waste management. Made from polyethylene, it features a stackable design and ergonomic handles for easy emptying. It is also NSF certified for food safety applications.
  • Made of durable plastic, reinforced with drag skids for maximum longevity.
  • Stackable design prevents vacuum seal during transport.
  • Ergonomic hand holds on the bottom facilitate easy emptying.
  • Integrated bag cinches secure liners.
  • Food safe and NSF 2 and NSF 21 certified for storing food products.
  • Color code program aligns with HACCP practices for sanitary maintenance.
  • Made in the USA.
  • Recyclable (LDPE, code 4).