Learn More
MSC Carlisle Trash Container, 20 gal, Round, Polyethylene, Gray
Catalog No. 502839232
Encompass
The Carlisle Bronco 20 gal Round Trash Container is a durable polyethylene waste bin designed for commercial establishments. It is suitable for both waste and food storage and meets HACCP, NSF 2, and NSF 21 standards.
  • Designed for commercial establishments
  • Durable polyethylene construction
  • Suitable for both waste and food storage
  • Gray color enables color-coded systems
  • Meets HACCP, NSF 2, and NSF 21 standards
Catalog No. 50-283-9232 Supplier MSC Supplier No. 73995656
The Carlisle Bronco 20 gal Round Trash Container is a durable polyethylene waste bin designed for commercial establishments. It is suitable for both waste and food storage and meets HACCP, NSF 2, and NSF 21 standards.
  • Designed for commercial establishments
  • Durable polyethylene construction
  • Suitable for both waste and food storage
  • Gray color enables color-coded systems
  • Meets HACCP, NSF 2, and NSF 21 standards