Learn More
MSC Carlisle Round Waste Bin Trash Container, 20 Gallon, Red, Polyethylene
Catalog No. 502839261
Encompass
These durable round waste bins are designed for efficient waste management. Each 20 gallon container is constructed from polyethylene, featuring reinforced drag skids, a stackable design, and ergonomic hand holds for easy emptying. They are NSF 2 and NSF 21 certified, making them suitable for food handling environments.
  • Durable plastic with reinforced drag skids for longevity.
  • Stackable design prevents vacuum seal during transport.
  • Ergonomic hand holds for easy and efficient emptying.
  • Integrated bag cinches.
  • NSF 2 and NSF 21 certified for food safety and color-coding.
  • Aligns with HACCP practices for sanitary maintenance.
  • Made in the USA.
  • Recyclable (low-density polyethylene).
  • Available in multiple colors.
Catalog No. 50-283-9261 Supplier MSC Supplier No. 73994816
These durable round waste bins are designed for efficient waste management. Each 20 gallon container is constructed from polyethylene, featuring reinforced drag skids, a stackable design, and ergonomic hand holds for easy emptying. They are NSF 2 and NSF 21 certified, making them suitable for food handling environments.
  • Durable plastic with reinforced drag skids for longevity.
  • Stackable design prevents vacuum seal during transport.
  • Ergonomic hand holds for easy and efficient emptying.
  • Integrated bag cinches.
  • NSF 2 and NSF 21 certified for food safety and color-coding.
  • Aligns with HACCP practices for sanitary maintenance.
  • Made in the USA.
  • Recyclable (low-density polyethylene).
  • Available in multiple colors.