Learn More
MSC Carlisle Round Waste Bin Trash Container, 20 Gallon, Polyethylene, Green
Catalog No. 502839190
Encompass
This round waste bin trash container is constructed from durable polyethylene (LLDPE) and features reinforced drag skids, ensuring maximum longevity. With a 20 gallon capacity, it is designed for versatile use, including food storage, as it is NSF 2 and NSF 21 certified. The integrated color code program supports efficient sanitary maintenance practices.
  • Reinforced with drag skids for maximum longevity.
  • Stackable design prevents vacuum seal during transport.
  • Ergonomic handholds on the bottom for easy emptying.
  • Integrated bag cinches for secure liner attachment.
  • Food safe, NSF 2 and NSF 21 certified.
  • Color code program aligns with HACCP practices.
  • Made in the USA.
  • Recyclable, made from low-density polyethylene.
Catalog No. 50-283-9190 Supplier MSC Supplier No. 73995649
This round waste bin trash container is constructed from durable polyethylene (LLDPE) and features reinforced drag skids, ensuring maximum longevity. With a 20 gallon capacity, it is designed for versatile use, including food storage, as it is NSF 2 and NSF 21 certified. The integrated color code program supports efficient sanitary maintenance practices.
  • Reinforced with drag skids for maximum longevity.
  • Stackable design prevents vacuum seal during transport.
  • Ergonomic handholds on the bottom for easy emptying.
  • Integrated bag cinches for secure liner attachment.
  • Food safe, NSF 2 and NSF 21 certified.
  • Color code program aligns with HACCP practices.
  • Made in the USA.
  • Recyclable, made from low-density polyethylene.