Learn More
MSC Carlisle Waste Bin Trash Container, Round, 55 Gallon, Polyethylene, Brown
Catalog No. 502836019
Encompass
This round waste bin is crafted from durable polyethylene, featuring reinforced drag skids for longevity and easy movement. Its stackable design prevents vacuum seals during transport, and ergonomic hand holds facilitate efficient emptying. Integrated bag cinches add convenience, and the container is food-safe (NSF 2 and NSF 21 certified), supporting HACCP color-coded sanitary systems.
  • Durable polyethylene construction with reinforced drag skids.
  • Stackable design prevents vacuum seal during transport.
  • Ergonomic bottom hand holds for easy emptying.
  • Integrated bag cinches.
  • Food safe, NSF 2 and NSF 21 certified.
  • Aligns with HACCP practices for color-coded sanitary systems.
Catalog No. 50-283-6019 Supplier MSC Supplier No. 73997942
This round waste bin is crafted from durable polyethylene, featuring reinforced drag skids for longevity and easy movement. Its stackable design prevents vacuum seals during transport, and ergonomic hand holds facilitate efficient emptying. Integrated bag cinches add convenience, and the container is food-safe (NSF 2 and NSF 21 certified), supporting HACCP color-coded sanitary systems.
  • Durable polyethylene construction with reinforced drag skids.
  • Stackable design prevents vacuum seal during transport.
  • Ergonomic bottom hand holds for easy emptying.
  • Integrated bag cinches.
  • Food safe, NSF 2 and NSF 21 certified.
  • Aligns with HACCP practices for color-coded sanitary systems.