Learn More
Apexbio Technology LLC BRONOPOL-100MG
SDP
Catalog No. 5000570013
Encompass_Preferred
Bronopol (CAS No 52-51-7) is a biocidal compound primarily used in biomedical research for its antimicrobial properties Originating as a preservative its mechanism of action involves disrupting bacterial protein synthesis by releasing formaldehyde targeting thiol groups in proteins In vitro studies typically observe activity in the nanomolar to low micromolar range Common applications include testing in cell models such as bacterial cultures as well as animal studies for evaluating microbial inhibition It is used in drug screening for developing disinfectants and antimicrobial agents Experimental concentrations typically depend on the specific research design
Catalog No. 50-005-70013 Supplier Apexbio Technology LLC Supplier No. C6769100
Bronopol (CAS No 52-51-7) is a biocidal compound primarily used in biomedical research for its antimicrobial properties Originating as a preservative its mechanism of action involves disrupting bacterial protein synthesis by releasing formaldehyde targeting thiol groups in proteins In vitro studies typically observe activity in the nanomolar to low micromolar range Common applications include testing in cell models such as bacterial cultures as well as animal studies for evaluating microbial inhibition It is used in drug screening for developing disinfectants and antimicrobial agents Experimental concentrations typically depend on the specific research design