Learn More
Medchemexpress LLC Bronopol | 52-51-7 | 98.0% | 1 ML
SDP
Catalog No. 5000411302
Encompass_Preferred
Bronopol is an antibacterial agent with low toxicity to mammals and high activity, especially against Gram-negative bacteria. It functions by oxidizing protein thiols, inhibiting enzymatic activity, and also acts as a formaldehyde releaser. This solution is provided as 10 mM in 1 mL of DMSO, ready for research applications.
  • Antibacterial agent
  • Low toxicity to mammals
  • High activity against Gram-negative bacteria
  • Oxidizes protein thiols
  • Inhibits enzymatic activity
  • Formaldehyde releaser
Catalog No. 50-004-11302 Supplier Medchemexpress LLC Supplier No. HYB121710MM/1ML
Bronopol is an antibacterial agent with low toxicity to mammals and high activity, especially against Gram-negative bacteria. It functions by oxidizing protein thiols, inhibiting enzymatic activity, and also acts as a formaldehyde releaser. This solution is provided as 10 mM in 1 mL of DMSO, ready for research applications.
  • Antibacterial agent
  • Low toxicity to mammals
  • High activity against Gram-negative bacteria
  • Oxidizes protein thiols
  • Inhibits enzymatic activity
  • Formaldehyde releaser