Learn More
Medchemexpress LLC Bronopol | 52-51-7 | 98.0% | 100 G
SDP
Supplier:  Medchemexpress LLC HYB1217100G
Encompass_Preferred
Bronopol is an antibacterial agent effective against Gram-negative bacteria. It functions by oxidizing protein thiols, inhibiting enzymatic activity, and acting as a formaldehyde releaser. In vitro, it activates THP-1 cells, increases CD86 expression, induces IL-8 secretion, and promotes early expression of HMOX1 and HSPA6 genes. In vivo, it can induce chemical peritonitis and transitory pain aggressive behavior in Sprague-Dawley rat models.
  • Antibacterial agent with low toxicity to mammals
  • Effective against Gram-negative bacteria
  • Oxidizes protein thiols and inhibits enzymatic activity
  • Acts as a formaldehyde releaser
  • Activates THP-1 cells and increases CD86 expression in vitro
  • Induces IL-8 secretion and promotes gene expression in vitro
  • Can induce chemical peritonitis and pain in Sprague-Dawley rat models in vivo
Catalog No. 50-004-11149