Learn More
Medchemexpress LLC Bronopol | 52-51-7 | 98.0% | 500 G
SDP
Catalog No. 5000413652
Encompass_Preferred
Bronopol is an antibacterial agent with low toxicity and high activity, particularly against Gram-negative bacteria. It oxidizes protein thiols, inhibits enzymatic activity, and acts as a formaldehyde releaser. It is suitable for laboratory research, including in vitro and in vivo studies.
  • Oxidizes protein thiols
  • Inhibits enzymatic activity
  • Exhibits antibacterial activity
  • Functions as a formaldehyde releaser
  • Activates THP-1 cells
  • Increases CD86 expression
  • Induces IL-8 secretion
  • Promotes early expression of HMOX1 and HSPA6 genes
Catalog No. 50-004-13652 Supplier Medchemexpress LLC Supplier No. HYB1217500G
Bronopol is an antibacterial agent with low toxicity and high activity, particularly against Gram-negative bacteria. It oxidizes protein thiols, inhibits enzymatic activity, and acts as a formaldehyde releaser. It is suitable for laboratory research, including in vitro and in vivo studies.
  • Oxidizes protein thiols
  • Inhibits enzymatic activity
  • Exhibits antibacterial activity
  • Functions as a formaldehyde releaser
  • Activates THP-1 cells
  • Increases CD86 expression
  • Induces IL-8 secretion
  • Promotes early expression of HMOX1 and HSPA6 genes