Learn More
Electron Microscopy Sciences Broken Cross Line Reticle RET 50, 19 mm, 13 µm Line width
SDP
Catalog No. 501901745
Encompass_Preferred
High quality, precision made standard reticles are manufactured to use with our microscope eyepieces. All recticles are chrome on glass and positive polarity.
Size: 19 mm
Line width: 13 µm
Catalog No. 50-190-1745 Supplier Electron Microscopy Sciences Supplier No. 6025519
High quality, precision made standard reticles are manufactured to use with our microscope eyepieces. All recticles are chrome on glass and positive polarity.
Size: 19 mm
Line width: 13 µm