Learn More
Brookfield Engineer Labs Brookfield Ametek RV-2 Spindle, 302 stainless steel, Range-100-80K
Catalog No. NC1184211
Encompass
All standard RV, HA or HB torque Brookfield Viscometers/Rheometers are supplied with a standard spindle set (RV-2 through RV-7) constructed of #302 stainless steel. Additional spindle options are available in #316 stainless steel or Teflon coating for increased corrosion resistance. An optional spindle rack makes a convenient storage "bench" and allows easy and quick access to all spindles.
Catalog No. NC1184211 Supplier Brookfield Engineer Labs Supplier No. RV2
All standard RV, HA or HB torque Brookfield Viscometers/Rheometers are supplied with a standard spindle set (RV-2 through RV-7) constructed of #302 stainless steel. Additional spindle options are available in #316 stainless steel or Teflon coating for increased corrosion resistance. An optional spindle rack makes a convenient storage "bench" and allows easy and quick access to all spindles.