Learn More
MSC Vikan Handle, Stainless Steel, 59.4 in, Euro Threaded, Yellow
Catalog No. 502838428
Encompass
This tough and chemical-resistant stainless steel handle is ergonomically designed for greater comfort and durability. It is suitable for demanding use in hygiene-critical environments like food processing, retail, and service, and can be used with various compatible tools.
  • Designed for hygiene and food safety critical environments.
  • Withstands harsh chemicals and cleaning agents.
  • Durable construction for long-lasting performance.
  • Ergonomic design enhances comfort and reduces strain.
  • Maintains hygiene through easy cleaning and color-coding.
  • Drop-shaped hanging hole prevents liquid pooling for easy storage.
  • Euro threading ensures secure tool attachment.
  • Meets food contact material regulations: (EC) 1935/2004, FDA CFR 21, UK 2019 No. 704.
Catalog No. 50-283-8428 Supplier MSC Supplier No. 51879856
This tough and chemical-resistant stainless steel handle is ergonomically designed for greater comfort and durability. It is suitable for demanding use in hygiene-critical environments like food processing, retail, and service, and can be used with various compatible tools.
  • Designed for hygiene and food safety critical environments.
  • Withstands harsh chemicals and cleaning agents.
  • Durable construction for long-lasting performance.
  • Ergonomic design enhances comfort and reduces strain.
  • Maintains hygiene through easy cleaning and color-coding.
  • Drop-shaped hanging hole prevents liquid pooling for easy storage.
  • Euro threading ensures secure tool attachment.
  • Meets food contact material regulations: (EC) 1935/2004, FDA CFR 21, UK 2019 No. 704.