Learn More
MSC Vikan Stainless Steel Handle, 31 mm Diameter, 1025 mm Length, Green
Catalog No. 502839566
Encompass
This stainless steel handle is designed for use in environments requiring high hygiene, including those with aggressive chemicals. It pairs with various tools.
  • Purpose-built for food manufacturing, retail, restaurants, and food service where hygiene and food safety are critical.
  • Withstands harsh chemicals and cleaning agents.
  • Ergonomic vertical ridges and a matte finish provide a better grip, even with wet or greasy hands.
  • High-strength, corrosion-resistant stainless steel ensures durability for tough jobs.
  • Durable construction provides long-lasting performance.
  • Ergonomic design enhances comfort and reduces worker strain.
  • Easy to clean and maintain for hygiene control.
  • Color-coded for hygienic zoning plans.
  • Euro threading ensures secure tool attachment.
  • Drop-shaped hanging hole prevents liquid pooling and eases storage.
Catalog No. 50-283-9566 Supplier MSC Supplier No. 84850759
This stainless steel handle is designed for use in environments requiring high hygiene, including those with aggressive chemicals. It pairs with various tools.
  • Purpose-built for food manufacturing, retail, restaurants, and food service where hygiene and food safety are critical.
  • Withstands harsh chemicals and cleaning agents.
  • Ergonomic vertical ridges and a matte finish provide a better grip, even with wet or greasy hands.
  • High-strength, corrosion-resistant stainless steel ensures durability for tough jobs.
  • Durable construction provides long-lasting performance.
  • Ergonomic design enhances comfort and reduces worker strain.
  • Easy to clean and maintain for hygiene control.
  • Color-coded for hygienic zoning plans.
  • Euro threading ensures secure tool attachment.
  • Drop-shaped hanging hole prevents liquid pooling and eases storage.