Learn More
Cayman Chemical Broprmn 10mg
SDP
Catalog No. 502990210
Encompass_Preferred
An immunomodulator with diverse biological activities; a TLR7 agonist that inhibits RANKL-induced osteoclast differentiation of murine BMDMs; dietary administration prevents development of TCCs in a mouse model of carcinogen-induced bladder carcinoma at 200 mg/kg twice per week; reduces pulmonary collagen accumulation and BALF monocyte and eosinophil infiltration in a hamster model of bleomycin-induced lung fibrosis at 100 mg/kg; delays disease onset and progression and increases plasma IFN levels in a mouse model of EAE
Catalog No. 50-299-0210 Supplier Cayman Chemical Supplier No. 2944010 MG
An immunomodulator with diverse biological activities; a TLR7 agonist that inhibits RANKL-induced osteoclast differentiation of murine BMDMs; dietary administration prevents development of TCCs in a mouse model of carcinogen-induced bladder carcinoma at 200 mg/kg twice per week; reduces pulmonary collagen accumulation and BALF monocyte and eosinophil infiltration in a hamster model of bleomycin-induced lung fibrosis at 100 mg/kg; delays disease onset and progression and increases plasma IFN levels in a mouse model of EAE