Learn More
Enterprise Technology Solutions Brother Label Maker,Bluetooth&USB,f/24mm TzeLamTapes,Customize,Apple&Android,w/StarterTape,5"x2.6"x5",BK
SDP
Catalog No. 5000740212
Encompass_Preferred
Design and print custom labels with the P-touch CUBE Plus Label Maker. Bluetooth wireless technology allows you to wirelessly print labels up to 24mm (~1") wide from your smartphone, tablet or laptop, and it's compatible with both Apple and Android devices for ultimate convenience. The provided USB cable also let you print from a desktop. Choose from three different applications to print the custom labels you need: the Brother iPrint&Label app, the P-touch Design&Print2 app or the P-touch Editor software from your PC. Templates include 450 symbols, over 60 frames and a variety of fonts to choose from. This label maker is compatible with Brother P-touch TZe durable laminated tapes. The CUBE comes with one 24mm (0.94") x 4m (13.1') Black Print on White Starter Label Tape and one 24mm (0.94") x 2m (6.5') Black Print on Premium Gold Starter Label Tape.
Catalog No. 50-007-40212 Supplier Enterprise Technology Solutions Supplier No. BRTPTP710BT
Design and print custom labels with the P-touch CUBE Plus Label Maker. Bluetooth wireless technology allows you to wirelessly print labels up to 24mm (~1") wide from your smartphone, tablet or laptop, and it's compatible with both Apple and Android devices for ultimate convenience. The provided USB cable also let you print from a desktop. Choose from three different applications to print the custom labels you need: the Brother iPrint&Label app, the P-touch Design&Print2 app or the P-touch Editor software from your PC. Templates include 450 symbols, over 60 frames and a variety of fonts to choose from. This label maker is compatible with Brother P-touch TZe durable laminated tapes. The CUBE comes with one 24mm (0.94") x 4m (13.1') Black Print on White Starter Label Tape and one 24mm (0.94") x 2m (6.5') Black Print on Premium Gold Starter Label Tape.