Learn More
Enterprise Technology Solutions Brother Label Maker,LCD,30mm/Sec,8"x7.7"x3.4",BK
SDP
Catalog No. 5000739560
Encompass_Preferred
Business Professional Connected Label Maker features high-resolution printing for sharper bar codes and more with full-color LCD screen. Create labels using your computer or mobile phone via Bluetooth with P-touch Editor Software1 and Brother iPrint app. Advanced connectivity allows multiple users with their own device to work with the same label maker. Increase efficiency on high-volume labeling jobs with faster printing and built-in automatic label cutter. Its maximum printing speed is 30mm per second. Enjoy sharp, vivid 180 x 360 dpi resolution. Easily type using the QWERTY keyboard, one-touch formatting keys and large 20-character, two-line screen preview. Choose between 17 fonts, 17 font styles, 160 frames, 900 symbols and 175 pre-set label templates. Increased memory capacity saves up to 90 used labels. It prints up to 99 copies. Labelmaker uses 1" tape and is compatible with Brother Genuine TZe Label Tapes for indoor and outdoor label versatility.
Catalog No. 50-007-39560 Supplier Enterprise Technology Solutions Supplier No. BRTPTD610BT
Business Professional Connected Label Maker features high-resolution printing for sharper bar codes and more with full-color LCD screen. Create labels using your computer or mobile phone via Bluetooth with P-touch Editor Software1 and Brother iPrint app. Advanced connectivity allows multiple users with their own device to work with the same label maker. Increase efficiency on high-volume labeling jobs with faster printing and built-in automatic label cutter. Its maximum printing speed is 30mm per second. Enjoy sharp, vivid 180 x 360 dpi resolution. Easily type using the QWERTY keyboard, one-touch formatting keys and large 20-character, two-line screen preview. Choose between 17 fonts, 17 font styles, 160 frames, 900 symbols and 175 pre-set label templates. Increased memory capacity saves up to 90 used labels. It prints up to 99 copies. Labelmaker uses 1" tape and is compatible with Brother Genuine TZe Label Tapes for indoor and outdoor label versatility.