Learn More
Enterprise Technology Solutions Brother Label Maker, PC/Mac, 3-1/5"x6"x5-3/5", Black/White
SDP
Catalog No. 5000739607
Encompass_Preferred
PC-connectable label maker offers simple connectivity for your PC and Mac with no software or drivers to install. Design custom labels using fonts and graphics from your PC or Mac and the included label design software. Download templates from the Cloud to add to your label library (Internet connection required). With super-fast printing and auto cutter, generate extra-wide, laminated TZe tapes - 9/64", 1/4", 3/8", 1/2", 3/4", 1" (3.5mm, 6mm, 9mm, 12mm, 18mm, 24mm) widths for indoor/outdoor use. Formatting options include logos, bar codes, frames and symbols on any label. The PT-P700 includes an AC adapter, USB cable, software and starter tape.
Catalog No. 50-007-39607 Supplier Enterprise Technology Solutions Supplier No. BRTPTP700
PC-connectable label maker offers simple connectivity for your PC and Mac with no software or drivers to install. Design custom labels using fonts and graphics from your PC or Mac and the included label design software. Download templates from the Cloud to add to your label library (Internet connection required). With super-fast printing and auto cutter, generate extra-wide, laminated TZe tapes - 9/64", 1/4", 3/8", 1/2", 3/4", 1" (3.5mm, 6mm, 9mm, 12mm, 18mm, 24mm) widths for indoor/outdoor use. Formatting options include logos, bar codes, frames and symbols on any label. The PT-P700 includes an AC adapter, USB cable, software and starter tape.