Learn More
Enterprise Technology Solutions Brother Label Maker,w/Case,20mm/sec,3/4"Labels,7.4"x3.9"12.3",WE
Supplier: Enterprise Technology Solutions BRTPTD410VP
Home/Office Advanced Label Maker Value Pack is a versatile, easy-to-use way to organize your home or office. Connect it to a PC or Mac with included USB cable and create labels using P-touch Editor Software1. Simply use the QWERTY keyboard and quick, one-touch formatting keys to quickly print your labels. Organize the contents of your desk and more using up to 3/4" thick labels. It is compatible with 12mm, 9mm, 6mm, 3.5mm and 18mm tapes. Use Genuine Brother TZe Label Tapes for indoor and outdoor versatility. Select between 15 fonts, 13 font styles, 120 frames, 700 symbols and 30 pre-set label templates for professional labels. Preview labels to avoid errors before printing with the 15-character, two-line display screen. Labels print with sharp, 180 dpi print resolution. Print labels up to 20mm per second. Memory bank saves up to 50 frequently used labels. Power this desktop labelmaker using AC adapter (included) or six AAA batteries (not included). Easily carry using the included case.