Learn More
Enterprise Technology Solutions Brother Label Tape,Flexible,Lamin,f/P-touch,0.7"x26.2',BlackonWhite
SDP
Catalog No. 5000739941
Encompass_Preferred
Specifically designed to wrap securely around curved surfaces and withstand extreme conditions, Genuine TZe Flexible ID Laminated Label Tape helps you organize your wires, cables, pipes and so much more. A reliable choice for both indoor and outdoor applications, this strong, durable tape can be used confidently in harsh environments and on rough/uneven surfaces. Like all Brother P-touch TZe Label Tapes, it is designed to work seamlessly with variety of Brother P-touch Label Makers and engineered for effortless customization, long-lasting performance and impressive durability. Flexible tape is chemical-resistant, water-resistant, abrasion-resistant and fade-resistant so it is great for use in medical offices, nursing homes, janitorial supply areas, gyms, locker rooms, auto shops, kitchens, boating, gardening supply centers. Print is black. Background is white.
Catalog No. 50-007-39941 Supplier Enterprise Technology Solutions Supplier No. BRTTZEFX241
Specifically designed to wrap securely around curved surfaces and withstand extreme conditions, Genuine TZe Flexible ID Laminated Label Tape helps you organize your wires, cables, pipes and so much more. A reliable choice for both indoor and outdoor applications, this strong, durable tape can be used confidently in harsh environments and on rough/uneven surfaces. Like all Brother P-touch TZe Label Tapes, it is designed to work seamlessly with variety of Brother P-touch Label Makers and engineered for effortless customization, long-lasting performance and impressive durability. Flexible tape is chemical-resistant, water-resistant, abrasion-resistant and fade-resistant so it is great for use in medical offices, nursing homes, janitorial supply areas, gyms, locker rooms, auto shops, kitchens, boating, gardening supply centers. Print is black. Background is white.