Learn More
Enterprise Technology Solutions Brother Label Tape,Xtra-Strength,f/P-touch,0.47"x26.2',WhiteonClear
SDP
Supplier:  Enterprise Technology Solutions BRTTZES135G
Encompass_Preferred
With twice the adhesive of Brother's standard TZe label tapes, this Genuine TZe Extra-Strength Laminated Label Tape is tough enough for almost any job. Apply with confidence knowing your label will stick there and stay there. A reliable choice for indoor and outdoor applications, confidently use the strong, durable tape in harsh environments and on rough/uneven surfaces. Like all P-touch TZe Label Tapes, it is designed to work seamlessly with variety of Brother P-touch Label Makers and engineered for effortless customization, long-lasting performance and impressive durability. Tape resists abrasion, water, many different chemicals and fading caused by sunlight. Use anywhere that needs extra sticking power like warehouses, schools, stockrooms, production floors and heavy equipment. Label tape is easy to apply and remains affixed to smooth, round and textured surfaces. Print color is white. Background is clear. Sustainable packaging is made with responsibly sourced FSC-certified paper.
Catalog No. 50-007-39837