Learn More
Enterprise Technology Solutions Brother Labeling Tape, Nonlaminated, 1/2" Size, 3/BD, Black/Blue
SDP
Catalog No. 5000740105
Encompass_Preferred
Create labels for home, home office, school or hobby applications using this 1/2" Black on Blue tape. It is designed for use with Brother P-Touch machines including PT-100, PT-110, PT-45M, PT-55BM, PT-55S, PT-65, PT-65SB, PT-65SCCP, PT-65SL, PT-65VP, PT-70, PT-70BBVP, PT-70BM, PT-70BMH, PT-70HK, PT-70HOL, PT-70HOT, PT-70SP, PT-70SR, PT-80, PT-80SCCP, PT-85, PT-90 and PT-M95. Nonlaminated tape is perfect for indoor and light use. It yields 26.2'.
Catalog No. 50-007-40105 Supplier Enterprise Technology Solutions Supplier No. BRTM531BD
Create labels for home, home office, school or hobby applications using this 1/2" Black on Blue tape. It is designed for use with Brother P-Touch machines including PT-100, PT-110, PT-45M, PT-55BM, PT-55S, PT-65, PT-65SB, PT-65SCCP, PT-65SL, PT-65VP, PT-70, PT-70BBVP, PT-70BM, PT-70BMH, PT-70HK, PT-70HOL, PT-70HOT, PT-70SP, PT-70SR, PT-80, PT-80SCCP, PT-85, PT-90 and PT-M95. Nonlaminated tape is perfect for indoor and light use. It yields 26.2'.