Learn More
Enterprise Technology Solutions Brother LabelMaker, P-Touch, 12-1/5"x7-1/2"x6-1/10", BK
SDP
Catalog No. 5000739561
Encompass_Preferred
Portable labelmaker lets you quickly create professional-style labels with fast print speeds up to 60mm per second. Easy-view backlit display shows you how your label will look before you print to prevent errors and minimize waste. Removable keyboard utilizes a familiar computer-style layout for quick, smooth input. Cable label wizard helps you easily organize and label your wires and cables. Plus, print from your mobile device with Brother applications. Integrated storage space provides a convenient location to keep your accessories readily available. Flexible design can be powered by the included AC adapter or rechargeable battery (sold separately) for on-the-go performance. Labelmaker is designed for use with Brother TZe tapes up to 36mm wide.
Catalog No. 50-007-39561 Supplier Enterprise Technology Solutions Supplier No. BRTPTD800W
Portable labelmaker lets you quickly create professional-style labels with fast print speeds up to 60mm per second. Easy-view backlit display shows you how your label will look before you print to prevent errors and minimize waste. Removable keyboard utilizes a familiar computer-style layout for quick, smooth input. Cable label wizard helps you easily organize and label your wires and cables. Plus, print from your mobile device with Brother applications. Integrated storage space provides a convenient location to keep your accessories readily available. Flexible design can be powered by the included AC adapter or rechargeable battery (sold separately) for on-the-go performance. Labelmaker is designed for use with Brother TZe tapes up to 36mm wide.