Learn More
Enterprise Technology Solutions Brother Tape Cartridge, 1/2" Size, 2/PK, Black/Clear
SDP
Catalog No. 5000740213
Encompass_Preferred
Tape cartridges feature standard adhesive designed for flat surfaces like office paper, file folders and binders. Lamination resists water, grease and grime. Use in hot and cold environments as well as outdoors. Tape cartridges are designed for use with Brother PT-6, 8, 10, 12, 12N, 15, 20, 25, 150 and 170K labelers.
Catalog No. 50-007-40213 Supplier Enterprise Technology Solutions Supplier No. BRTTC10
Tape cartridges feature standard adhesive designed for flat surfaces like office paper, file folders and binders. Lamination resists water, grease and grime. Use in hot and cold environments as well as outdoors. Tape cartridges are designed for use with Brother PT-6, 8, 10, 12, 12N, 15, 20, 25, 150 and 170K labelers.