Learn More
Enterprise Technology Solutions Brother Tape Cartridge, 1/2" Size, 2/PK, Red/White
SDP
Catalog No. 5000740199
Encompass_Preferred
Tape cartridges feature standard adhesive designed for flat surfaces like office paper, file folders and binders. Lamination resists water, grease and grime. Use in hot and cold environments as well as outdoors. Tape cartridges are designed for use with Brother PT-6, 8, 10, 12, 12N, 15, 20, 25, 150 and 170K labelers.
Catalog No. 50-007-40199 Supplier Enterprise Technology Solutions Supplier No. BRTTC21
Tape cartridges feature standard adhesive designed for flat surfaces like office paper, file folders and binders. Lamination resists water, grease and grime. Use in hot and cold environments as well as outdoors. Tape cartridges are designed for use with Brother PT-6, 8, 10, 12, 12N, 15, 20, 25, 150 and 170K labelers.