Learn More
Enterprise Technology Solutions Brother Tape Cartridge, 3/8" Size, White/Clear
SDP
Catalog No. 5000739620
Encompass_Preferred
Tape cartridges feature standard adhesive designed for flat surfaces like office paper, file folders and binders. Lamination resists water, grease and grime. Use in hot and cold environments as well as outdoors. Tape cartridges are designed for use with Brother PT-6, 8, 10, 12, 12N, 15, 20, 25, 150 and 170K labelers.
Catalog No. 50-007-39620 Supplier Enterprise Technology Solutions Supplier No. BRTTC14Z1
Tape cartridges feature standard adhesive designed for flat surfaces like office paper, file folders and binders. Lamination resists water, grease and grime. Use in hot and cold environments as well as outdoors. Tape cartridges are designed for use with Brother PT-6, 8, 10, 12, 12N, 15, 20, 25, 150 and 170K labelers.