Learn More
TARGETMOL CHEMICALS INC BROTIANIDE 5MG
SDP
Catalog No. 502580401
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. Brotianide (BAY-VA4059) has anthelmintic activity and is highly potent against liver fluke and gastric fluke infestations and can be used to treat acute subacute and chronic liver fascioliasis infections in sheep. purity: 98%
Catalog No. 50-258-0401 Supplier TARGETMOL CHEMICALS INC Supplier No. T305965MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. Brotianide (BAY-VA4059) has anthelmintic activity and is highly potent against liver fluke and gastric fluke infestations and can be used to treat acute subacute and chronic liver fascioliasis infections in sheep. purity: 98%