Learn More
Medchemexpress LLC Broussonin A | 73731-87-0 | 1 MG
SDP
Catalog No. 5000421737
Encompass_Preferred
Broussonin A is a potent BChE inhibitor, with an IC50 of 4.16 μM. It is a diarylpropane natural product isolated from the bark of Broussonetia papyrifera after solid fermentation. This compound suppresses VEGF-A-stimulated endothelial cell proliferation and abrogates angiogenic responses, making it suitable for research in cardiovascular and cancer disease areas.
  • Potent BChE inhibitor (IC50 of 4.16 μM)
  • Isolated from Broussonetia papyrifera
  • Suppresses VEGF-A-stimulated endothelial cell proliferation
  • Abrogates VEGF-A-stimulated angiogenic responses including cell migration, invasion, and tube formation
  • Shows anti-adipogenic activity against 3T3L1 cells
  • Exhibits transactivation of human recombinant ERbeta and ERalpha
Catalog No. 50-004-21737 Supplier Medchemexpress LLC Supplier No. HYN29601MG
Broussonin A is a potent BChE inhibitor, with an IC50 of 4.16 μM. It is a diarylpropane natural product isolated from the bark of Broussonetia papyrifera after solid fermentation. This compound suppresses VEGF-A-stimulated endothelial cell proliferation and abrogates angiogenic responses, making it suitable for research in cardiovascular and cancer disease areas.
  • Potent BChE inhibitor (IC50 of 4.16 μM)
  • Isolated from Broussonetia papyrifera
  • Suppresses VEGF-A-stimulated endothelial cell proliferation
  • Abrogates VEGF-A-stimulated angiogenic responses including cell migration, invasion, and tube formation
  • Shows anti-adipogenic activity against 3T3L1 cells
  • Exhibits transactivation of human recombinant ERbeta and ERalpha