Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mopec™ Brown Adson Forceps
SDP

Catalog No. 22445603 Shop All Mopec Products
Change view
Click to view available options
Length (Metric):
121 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
22-445-603 121 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-445-603 Supplier Mopec™ Supplier No. AB092
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Adson forceps made from stainless steel.

  • Extra broad ribbed grip
  • Straight with 7 x 7 teeth tips

Specifications

Autoclavable Not Autoclavable
Color Silver
Material Stainless Steel
Disposable Reusable
Latex Free Yes
Length (English) 4.75 in.
Length (Metric) 121 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.