Learn More
Alimed Inc BROWN-ADSON TISSUE FORCEPS
SDP
Catalog No. NC1577419
Encompass
Straight. Side grasping. 7 x 7 teeth. 4-3/4"L.
This item is not returnable. View return policy
Catalog No. NC1577419 Supplier Alimed Inc Supplier No. 2970018490
Straight. Side grasping. 7 x 7 teeth. 4-3/4"L.