Learn More
Bioworld BROWN SEPAROPOREAGAROSE)
Catalog No. 50197918
Encompass
This item has a minimum order quantity of 3 per supplier requirements. BROWN SEPAROPORE (AGAROSE) 4B-CL: A dye-ligand affinity matrix consisting of Reactive Brown 10bound toCross-linked 4% agarose beads (Separopore 4B-CL). Specifications: Matrix: Cross-linked agarose beads, 4% Dye:Reactive Bown 10 E...
Catalog No. 50-197-918 Supplier Bioworld Supplier No. 201811001
This item has a minimum order quantity of 3 per supplier requirements. BROWN SEPAROPORE (AGAROSE) 4B-CL: A dye-ligand affinity matrix consisting of Reactive Brown 10bound toCross-linked 4% agarose beads (Separopore 4B-CL). Specifications: Matrix: Cross-linked agarose beads, 4% Dye:Reactive Bown 10 E...