Learn More
Bioworld BROWN SEPAROPOREAGAROSE)
Supplier:  Bioworld 201811002
Encompass
BROWN SEPAROPORE (AGAROSE) 4B-CL: A dye-ligand affinity matrix consisting of Reactive Brown 10bound toCross-linked 4% agarose beads (Separopore 4B-CL). Specifications: Matrix: Cross-linked agarose beads, 4% Dye:Reactive Bown 10 E...
Catalog No. 50-197-919