Learn More
Perkin Elmer US LLC Brownlee Aquapore RP-300 Column - 220 mm x 4.6 mm I.D.
Supplier:  Perkin Elmer US LLC 07110059
Encompass_Preferred
Recommended for the separation of large biomolecules, such as peptides and proteins.
Catalog No. 50-211-3557