Learn More
Perkin Elmer US LLC Brownlee Spheri-5 Amino Cartridge Column - 220 mm x 4.6 mm I.D.
Catalog No. 502113560
Encompass_Preferred
Recommended for the normal-phase separation of smaller, relatively polar compounds.
Catalog No. 50-211-3560 Supplier Perkin Elmer US LLC Supplier No. 07110041
Recommended for the normal-phase separation of smaller, relatively polar compounds.