Learn More
Perkin Elmer US LLC Brownlee Spheri-5 C8 Conventional Column - 250 mm x 4.6 mm I.D.
Supplier:  Perkin Elmer US LLC 07120012
Encompass_Preferred
The Brownlee Spheri C18 ODS column contains a small-pore, silica-based sorbent for separating small molecules. The C18 is one of the least retentive reverse-phase in this family. This conventional Brownlee column (non-cartridge) uses standard compression end fittings compatible with all HPLC instrumentation and only requires a holder when used with a NewGuard cartridge. To couple a conventional column to a NewGuard cartridge, a NewGuard holder (07150001) is necessary. End-Capping: Yes Carbon Load: 6% Interaction Mode: Reversed Phase pH Range: 2.5-8.0 Temperature Limit: 60 USP Code: L7
Catalog No. 50-211-3625
Only null left
Add to Cart
Request a Quote