Learn More
Perkin Elmer US LLC Brownlee Spheri-5 RP-18 Cartridge Column - 30 mm x 2.1 mm I.D., Pkg. 2
Catalog No. 502113541
Encompass_Preferred
Monofunctional bonded C18 phase for general purpose reversed phase small molecule applications.
Catalog No. 50-211-3541 Supplier Perkin Elmer US LLC Supplier No. 07110014
Add to Cart
Add to Cart
Monofunctional bonded C18 phase for general purpose reversed phase small molecule applications.