Learn More
Perkin Elmer US LLC Brownlee SPP C18 Column - 100 mm , 2.1 mm I.D.
Catalog No. 502103940
Encompass_Preferred
High purity, general purpose Octadecyl (C18) phase with wide selectivity. Recommended for the reverse phase separation of acidic, basic, or neutral compounds.
Catalog No. 50-210-3940 Supplier Perkin Elmer US LLC Supplier No. N9308404
High purity, general purpose Octadecyl (C18) phase with wide selectivity. Recommended for the reverse phase separation of acidic, basic, or neutral compounds.