Learn More
Perkin Elmer US LLC Brownlee SPP C18 Column - 75 mm x 4.6 mm I.D.
Catalog No. 502103956
Encompass_Preferred
High purity, general purpose Octadecyl (C18) phase with wide selectivity. Recommended for the reverse phase separation of acidic, basic, or neutral compounds.
Catalog No. 50-210-3956 Supplier Perkin Elmer US LLC Supplier No. N9308415
High purity, general purpose Octadecyl (C18) phase with wide selectivity. Recommended for the reverse phase separation of acidic, basic, or neutral compounds.