Learn More
Perkin Elmer US LLC Brownlee SPP RP-Amide Column - 20 mm x 2.1 mm I.D.
Supplier:  Perkin Elmer US LLC N9308497
Encompass_Preferred
Excellent phase for significantly increasing the retention and selectivity of acids, for symmetrical peak shapes of bases, zwitterions and other polar compounds.
Catalog No. 50-210-4042