Learn More
Medchemexpress LLC Broxaldine | 3684-46-6 | 99.88% | 250 MG
SDP
Catalog No. 5000414399
Encompass_Preferred
Broxaldine is an antiprotozoal agent that inhibits *Clostridium difficile* with a MIC value of 4 μM and exhibits antifungal effects. It is a solid with a white to off-white appearance. This product is for research use only and not intended for human use.
  • Inhibits *Clostridium difficile*
  • Exhibits antifungal effects
  • Supplied as a solid
  • Appearance is white to off-white
Catalog No. 50-004-14399 Supplier Medchemexpress LLC Supplier No. 250MGHYB1143
Broxaldine is an antiprotozoal agent that inhibits *Clostridium difficile* with a MIC value of 4 μM and exhibits antifungal effects. It is a solid with a white to off-white appearance. This product is for research use only and not intended for human use.
  • Inhibits *Clostridium difficile*
  • Exhibits antifungal effects
  • Supplied as a solid
  • Appearance is white to off-white