Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Broxaldine | 3684-46-6 | MFCD00865040 | 99.9% | 5 MG
SDP

Catalog No. 5000413731
Encompass_Preferred

Broxaldine (Brobenzoxaldine) is an antiprotozoal agent known to inhibit Clostridium difficile with a MIC value of 4 μM, and it also demonstrates antifungal effects. This product is for research use only.

  • Acts as an antiprotozoal agent
  • Inhibits Clostridium difficile
  • Exhibits antifungal properties
  • Purity of 99.88%
  • Supplied as a solid, white to off-white in appearance
  • Soluble in DMSO at 8.33 mg/mL
  • Specific storage conditions for powder and solutions are provided for stability

Catalog No. 50-004-13731 Supplier Medchemexpress LLC Supplier No. HYB11435MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Broxaldine (Brobenzoxaldine) is an antiprotozoal agent known to inhibit Clostridium difficile with a MIC value of 4 μM, and it also demonstrates antifungal effects. This product is for research use only.

  • Acts as an antiprotozoal agent
  • Inhibits Clostridium difficile
  • Exhibits antifungal properties
  • Purity of 99.88%
  • Supplied as a solid, white to off-white in appearance
  • Soluble in DMSO at 8.33 mg/mL
  • Specific storage conditions for powder and solutions are provided for stability

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.